Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moesin Antibody (MSN/491), Novus Biologicals™
SDP

Catalog No. NBP232875 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232875 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP232875 Supplier Novus Biologicals Supplier No. NBP232875
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Moesin Monoclonal specifically detects Moesin in Human, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Moesin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MSN/491
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 0.5-1 μg/mL, Simple Western 1.0 μg/mL, Flow Cytometry 0.5-1 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 0.5-1 μg/mL, Immunoprecipitation 0.5-1 μg/500 μg protein lysate, Immunohistochemistry-Paraffin 0.5-1.0 μg/mL, Immunohistochemistry-Frozen 0.5-1.0 μg/mL
Gene Accession No. P26038
Gene Alias Membrane-organizing extension spike protein, moesin
Gene Symbols MSN
Host Species Mouse
Immunogen Recombinant full-length human Moesin protein (Uniprot: P26038)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cytoskeleton Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4478
Test Specificity Recognizes 78kDa moesin protein. Moesin, a member of the talin-4.1 superfamily, is a linking protein of the sub-membranous actin cytoskeleton. It is expressed in variable amounts in cells of different phenotypes such as macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. The ERM proteins, ezrin, radixin, and moesin are involved in a variety of cellular functions, such as cell adhesion, migration, and the organization of cell surface structures, and are highly homologous, both in protein sequence and in functional activity, with merlin/schwannomin, a neurofibromatosis-2-associated tumor-suppressor protein. Cell lines of epithelial and mesothelial origin contain both moesin and radixin whereas cells of endothelial and lymphoid origin express moesin.
Target Species Human, Rat
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.