Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moesin Antibody (MSN/492), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N247916PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N247916PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N247916PEC5 Supplier Novus Biologicals Supplier No. NBP247916PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Moesin Monoclonal specifically detects Moesin in Human, Rat (Negative) samples. It is validated for Flow Cytometry, Knockout Validated.

Specifications

Antigen Moesin
Applications Flow Cytometry, Immunoassay
Classification Monoclonal
Clone MSN/492
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry, Knockout Validated
Formulation PBS with 0.05% Sodium Azide
Gene Alias Membrane-organizing extension spike protein, moesin
Gene Symbols MSN
Host Species Mouse
Immunogen Recombinant full-length human Moesin protein (Uniprot: P26038 )
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cytoskeleton Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4478.0
Test Specificity Recognizes 78kDa moesin protein. Moesin, a member of the talin-4.1 superfamily, is a linking protein of the sub-membranous actin cytoskeleton. It is expressed in variable amounts in cells of different phenotypes such as macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. The ERM proteins, ezrin, radixin, and moesin are involved in a variety of cellular functions, such as cell adhesion, migration, and the organization of cell surface structures, and are highly homologous, both in protein sequence and in functional activity, with merlin/schwannomin, a neurofibromatosis-2-associated tumor-suppressor protein. Cell lines of epithelial and mesothelial origin contain both moesin and radixin whereas cells of endothelial and lymphoid origin express moesin.
Target Species Human, Rat (Negative)
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.