Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moesin Antibody (rMSN/492), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB383601 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383601 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383601 Supplier Novus Biologicals Supplier No. NBP308923AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Moesin Monoclonal antibody specifically detects Moesin in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Moesin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rMSN/492
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias Membrane-organizing extension spike protein, moesin
Host Species Mouse
Immunogen Recombinant full-length human Moesin protein (Uniprot: P26038)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4478
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.