Learn More
Description
Specifications
Specifications
| Antigen | Moesin |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | SPM562 |
| Conjugate | DyLight 680 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated |
| Gene Alias | Membrane-organizing extension spike protein, moesin |
| Gene Symbols | MSN |
| Host Species | Mouse |
| Immunogen | Moesin Purified from uterus (Uniprot: P26038) |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
