Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mohawk homeobox Antibody (OTI6B12), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP27278A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27278A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27278A32 Supplier Novus Biologicals Supplier No. NBP272758AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mohawk homeobox Monoclonal specifically detects Mohawk homeobox in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Mohawk homeobox
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI6B12
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias C10orf48, homeobox protein Mohawk, IFRX, Iroquois family related homeodomain protein, iroquois homeobox protein-like 1, IRXL1chromosome 10 open reading frame 48, MGC39616, mohawk homeobox
Gene Symbols MKX
Host Species Mouse
Immunogen Full length recombinant protein of human Mohawk homeobox (NP_775847) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 283078
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.