Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFG Tray Stacking Box

Catalog No. p-4402988 Shop All Molded Fiber Glass Tray Company Products
Change view
Click to view available options
For Use With (Application):
Forging, Rubber Stamping
Material Handling and Storage
Product Type:
Lid
Storage Box
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Product Type
15-239-87 Forging, Rubber Stamping Storage Box
15-239-94 Material Handling and Storage Lid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-239-87 Supplier MFG Tray Supplier No. 8081085269
Only null left
Add to Cart
Add to Cart

Easily stackable boxes

  • For forging, machine parts, stamping, ceramic, rubber and other hot parts
  • Impervious to standard cutting, oil, grease and solvents
  • Box Capacity: 3.8L (1 gal.)
  • Color: White

Specifications

Capacity (English) 1 gal.
Closure Type Lid
Color White
Material Fiberglass
Shape Rectangular
For Use With (Application) Forging, Rubber Stamping
Mounting Type Stackable
Product Type Storage Box
Length (English) 22 in.
Width (English) 12 in.
Height (English) 1.25 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.