Learn More
3M™ Molecular Detection Matrix Control MDMC96, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. s26898 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
:
Molecular Detection Matrix Control MDMC96, Sold by AquaPhoenix Scientific
Catalog No.
S26898 Molecular Detection Matrix Control MDMC96, Sold by AquaPhoenix Scientific
1 options
Catalog No. S26898 Supplier 3M™ Supplier No. MDMC96NA
Kit contains 96 tests, for use with 3M™ Molecular Detection System to check for inhibition of matrix sample during validation periods, when adopting 3M™ method or testing new/unknown matrices
  • Verify new or unknown matrices not inhibitory to amplification
  • Ready-to-use and pre-dispensed reagents
  • Only two transfer steps after enrichment
  • Closed-tube system reduces risk of amplicon contamination
  • Designed to work with 3M™ proprietary enrichment broths to promote growth
Specifications
Product Type Microbiology Media
Test Type Environmental Test
For Use With (Application) When adopting 3M™ method or testing new/unknown matrices
For Use With (Equipment) 3M™ Molecular Detection System
Quantity 96 Pack