Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BacMam GFP Transduction Control (BacMam 2.0)

Catalog No. B10383
Encompass_Preferred
Click to view available options
Quantity:
1 mL

Includes

BacMam GFP transduction control contains 1mL of virus at a concentration of approximately 1 x 108 viral particles/mL. Approximate fluorescence excitation/emission maxima for emGFP: 485/520 in nm.

Important part of the enhanced BacMam technology

The BacMam 2.0 Transduction Control lets you experience the power and convenience of enhanced BacMam technology, with the convenient readout of emerald GFP (emGFP); our Aequorea victoria emGFP is extremely bright and photostable and this non-targeted form is ideal for testing transduction. Check out how well BacMam 2.0 works with GFP in your model before advancing to the more specialized reagents. The BacMam technology is based on an insect virus (baculovirus) to help efficiently deliver and express genes in mammalian cells. The baculovirus has been modified to include an expression cassette for transgene expression in mammalian cells. BacMam 2.0 incorporate elements that help greatly enhance transduction efficiency and expression levels: a pseudotyped capsid protein for more efficient cell entry and genetic elements (enhanced CMV promoter and Woodchuck Post-transcriptional Regulatory Element), that boost expression levels. Baculoviruses do not replicate in mammalian cells and thus have an excellent safety profile and lack cytopathic effects on cells. Cell types previously not compatible with the technology (neuronal cells, T-cells), or only poorly transduced (some stem cells, CHO) are now transduced quantitatively in a simple, one step process.

  • Efficient: highly efficient (>90%) transduction of a wide range of mammalian cell lines, including primary cells, stem cells and neuronal cells
  • Fast and convenient: Simply add the BacMam 2.0 together with cells to your cover slip or microtiter plate, incubate overnight and assay or store frozen, assay-ready cells for later use
  • Safe: non-replicating in mammalian cells, lack of observable cytopathic effect and biosafety level (BSL) 1 handling
  • Enables Flexiblity: co-transduce more than one BacMam reagent for multiplex experiments or co-localization studies and tightly control expression levels by just varying the dose

Cell Analysis, Cell-Based Reporter Assays, Cellular Imaging, Fluorescent Protein Assays (GFP)

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Content And Storage The BacMam GFP ttransduction control contains 1 ml of virus at a concentration of approximately 1 × 10^8 viral particles⁄ml.
Approximate fluorescence excitation⁄emission maxima for emGFP: 485⁄520 in nm.
The BacMam transduction control should be stored at 4°C, protected from light. When stored as directed, it is stable for 6 months. DO NOT FREEZE.
Excitation Wavelength Range 485⁄520
Dye Type GFP (EmGFP)
For Use With (Equipment) Fluorescence Microscope, Fluorometer, HTS Reader, High Content Instrument, Flow Cytometer
Quantity 1 mL
Detection Method Fluorescence
Emission Visible
Form Liquid
Promoter CMV
Shipping Condition Wet Ice
Technique Fluorescence Intensity
Label Type Fluorescent Dye
Product Type BacMam GFP Transduction Control
SubCellular Localization Cytoplasm, Nucleus
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.