Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Sieve, Activated, Type 3A, 8 to 12 Mesh, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. p-5391617
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Wide Mouth Amber Glass Bottle
Wide Mouth PVC Coated Glass Bottle

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 1344-01-0
Molecular Formula AlCaNaO4Si+2
Molecular Weight (g/mol) 182.13
InChI Key IDZYHGDLWGVHQM-UHFFFAOYSA-N
PubChem CID 44144394
IUPAC Name aluminum;calcium;sodium;silicate
SMILES [O-][Si]([O-])([O-])[O-].[Na+].[Al+3].[Ca+2]

Specifications

Quantity 500 g
Specific Gravity 1L = 2.10kg
Mesh Size 8 to 12 Mesh
Packaging Wide Mouth Amber Glass Bottle
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Molecular Sieve, Activated, Type 3A
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.