Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular sieves, 13X, 1-2mm (0.04-0.08in) dia. pellets

Catalog No. p-7080391
Click to view available options
Quantity:
1 kg
5 kg

CAS: 63231-69-6

Molecular sieves is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is preferred for static dehydration in closed liquid or gas systems, e.g., in packaging of drugs, electric components and perishable chemicals; water scavenging in printing and plastics systems and drying saturated hydrocarbon streams. Generally considered a universal drying agent in polar and nonpolar media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63231-69-6
MDL Number MFCD00131613
IUPAC Name Molecular sieves 13X

Specifications

Quantity 1 kg
Sensitivity Hygroscopic
Particle Size 1/16 in.
Physical Form 1/16in Pellets
Solubility Information Insoluble in water.
Chemical Name or Material Molecular sieves, 13X, 1 to 2mm (0.04 to 0.08 in.) dia. pellets
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.