Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular sieves 13X, 8 to 12 mesh, ACROS Organics™

Click to view available options
Quantity:
500g
5kg
Packaging:
Glass bottle
Plastic drum
63231-69-6
MFCD00131613
63231-69-6
MFCD00131613
Drying agent
Quantity 500g
Color Beige
Bulk Density 600g/L min.
Residual Water 1.5% max.
Water Adsorption 23.5% min.
Packaging Glass bottle
Physical Form Beads
Chemical Name or Material Molecular sieves 13X, 13X, 8 to 12 mesh