Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular sieves 3A, 4 to 8 mesh, ACROS Organics™

Click to view available options
Quantity:
500g
5kg
Packaging:
Glass bottle
Plastic drum
308080-99-1
AlNaO6Si2
202.135
MFCD00131613
URGAHOPLAPQHLN-UHFFFAOYSA-N
19758701
aluminum;sodium;dioxido(oxo)silane
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
308080-99-1
AlNaO6Si2
202.135
MFCD00131613
URGAHOPLAPQHLN-UHFFFAOYSA-N
19758701
aluminum;sodium;dioxido(oxo)silane
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
Drying agent
Quantity 500g
Color Beige
Bulk Density 700 to 800g/L
Water Adsorption 20% min. (50%rH, 20°C, 24h)
Residual Water 1.5% max. (550°C, 2 hrs.)
Packaging Glass bottle
Solubility Information Solubility in water: insoluble.
Physical Form Granules
Chemical Name or Material Molecular sieves 3A, 4 to 8 mesh