Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular sieves 5A, 8 to 12 mesh

Catalog No. p-44831
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Glass bottle
Plastic drum

CAS: 69912-79-4 | AlNaO6Si2 | 202.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69912-79-4
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.14
MDL Number MFCD00147627
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
SMILES [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O

Specifications

Color Beige to Gray or Brown
Quantity 500 g
Bulk Density 680 to 760g/L
Packaging Glass bottle
Water Adsorption 20% min. (50% rH, 20°C, 24 Hrs.)
Residual Water 1.5% max. (550°C, 2 hrs.)
Physical Form Granules
IUPAC Name Molecular sieves 5A
Chemical Name or Material Molecular sieves 5A, 5A, 8 to 12 mesh
Hazard Category HYGROSCOPIC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.