Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdenum silicide, Mo 61% min, typically 99.5% (metals basis)

Catalog No. p-7048817
Click to view available options
Quantity:
50 g
250 g

CAS: 12136-78-6 | MoSi2 | 152.12 g/mol

In electrical resistors, in protective coatings at high temperatures, and in engine parts in space vehiclesMolybdenum silicide is used in electric furnaces, glass, steel, electronics and ceramics industries. It finds application in heat treatment materials. It is also used in microelectronics as a contact material. Further, it serves as a dry lubricant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In electrical resistors, in protective coatings at high temperatures, and in engine parts in space vehicles
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12136-78-6
Molecular Formula MoSi2
Molecular Weight (g/mol) 152.12
MDL Number MFCD00151535
InChI Key YXTPWUNVHCYOSP-UHFFFAOYSA-N
Synonym molybdenum disilicide, molybdenum silicide, ms silicide, molybdenum-silicide, molybdenum silicide, mo5si3, molybdenum silicide sputtering target, 50.8mm 2.0in dia x 3.18mm 0.125in thick, molybdenum silicide sputtering target, 50.8mm 2.0in dia x 6.35mm 0.250in thick, molybdenum silicide sputtering target, 76.2mm 3.0in dia x 3.18mm 0.125in thick, molybdenum silicide sputtering target, 76.2mm 3.0in dia x 6.35mm 0.250in thick
PubChem CID 6336985
IUPAC Name bis($l^{2}-silanylidene)molybdenum
SMILES [Si]=[Mo]=[Si]

Specifications

Melting Point 1870°C to 2030°C
Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in acids and aqua regia,however,it is soluble in HF and HNO3Soluble in nitric acid and hydrofluoric acid; Insoluble in water most acids.
Formula Weight 152.13
Percent Purity 99.5% (typically)
Concentration or Composition (by Analyte or Components) Mo 61% min.
Odor Odorless
Density 6.31 g/mL
Assay (metals basis)
Chemical Name or Material Molybdenum silicide
Show More Show Less
EINECSNumber 235-231-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.