Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdovanadate Reagent, TRAACS, Ricca Chemical
SDP

Catalog No. 52085 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52085 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52085 Supplier Ricca Chemical Company Supplier No. 52085
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9 , 12054-85-2 , 7803-55-6
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Color Green-Yellow
CAS Min % 90.0, 5.82, 1.81, 0.05
CAS Max % 94.0, 6.05, 1.88, 0.05
Grade Laboratory
Packaging Cubitainer
pH <3
Quantity 20 L
Physical Form Liquid
Chemical Name or Material Molybdovanadate Reagent, TRAACS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.