Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MON1A Antibody, Novus Biologicals™
SDP

Catalog No. NBP152006 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152006 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152006 Supplier Novus Biologicals Supplier No. NBP152006
Add to Cart
Add to Cart

Goat Polyclonal Antibody

MON1A Polyclonal specifically detects MON1A in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen MON1A
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_115731.2
Gene Alias FLJ97088, MON1 homolog A (yeast), SAND1MGC13272, vacuolar fusion protein MON1 homolog A
Gene Symbols MON1A
Host Species Goat
Immunogen Peptide with sequence C-RPLKTIYYTGPNE corresponding to internal region according to NP_001135973.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84315
Test Specificity This antibody is expected to recognize isoforms a and b (NP_115731.2; NP_001135973.1). Amino acid numbering in name refers to sequence NP_115731.2.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.