Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bio-Serv™ Monkey Shine Mirrors

Click to view available options
For Use With (Equipment):
With chain assembly
Description
- Made of durable stainless steel
- Shiny on both sides
- Smoothed edges for safety
- Available without chain assembly for use inside the cage
- Autoclavable
- Cagewasher safe
Specifications
Specifications
| Product Type | Primate Toy |
| Color | Shiny Silver |
| Material | Stainless Steel |
| For Use With (Equipment) | With chain assembly |
| Diameter (English) Inside | 6 in. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction