Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monkeypox Virus A29 Antibody (0025), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB350557 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB350557 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB350557 Supplier Novus Biologicals Supplier No. NBP318813MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Monkeypox Virus A29 Monoclonal antibody specifically detects Monkeypox Virus A29 in Virus samples. It is validated for Western Blot, ELISA, ELISA

Specifications

Antigen Monkeypox Virus A29
Applications Western Blot, ELISA, ELISA
Classification Monoclonal
Clone 0025
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen This product is a recombinant monoclonal antibody made to recombinant Monkeypox Virus A29 Protein expressed from HEK293 cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.