Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monkeypox Virus A29L Antibody (0027), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB350618 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB350618 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB350618 Supplier Novus Biologicals Supplier No. NBP318270MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Monkeypox Virus A29L Monoclonal antibody specifically detects Monkeypox Virus A29L in Virus samples. It is validated for Western Blot, ELISA, ELISA (Matched Antibody Pair)

Specifications

Antigen Monkeypox Virus A29L
Applications Western Blot, ELISA, ELISA Capture (Matched Antibody Pair)
Classification Monoclonal
Clone 0027
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, ELISA Detection (Matched Antibody Pair)
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Recombinant Monkeypox Virus A29L Protein.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.