Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monkeypox Virus B21R Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB292722 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292722 20 μL
NB292723 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB292722 Supplier Novus Biologicals Supplier No. NBP32610420UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Monkeypox Virus B21R Polyclonal antibody specifically detects Monkeypox Virus B21R in Virus samples. It is validated for Western Blot

Specifications

Antigen Monkeypox Virus B21R
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:20000
Formulation PBS, pH 7.0
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Monkeypox Virus B21R
Purification Method Antigen and protein A Affinity-purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.