Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monkeypox Virus B6R Antibody (004) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB306076 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB306076 20 μL
NB306077 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB306076 Supplier Novus Biologicals Supplier No. NBP32352520UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Monkeypox Virus B6R Monoclonal antibody specifically detects Monkeypox Virus B6R in Virus samples. It is validated for Western Blot, ELISA

Specifications

Antigen Monkeypox Virus B6R
Applications Western Blot, ELISA
Classification Monoclonal
Clone 004
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Monkeypox Virus B6R (Uniprot#: QNP13760.1; Tyr18-His279).
Purification Method Protein A purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.