Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mono- and polyubiquitinylated conjugates Antibody (UBCJ2), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB180047 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB180047 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB180047 Supplier Novus Biologicals Supplier No. NBP306879AF405
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mono- and polyubiquitinylated conjugates Monoclonal antibody specifically detects Mono- and polyubiquitinylated conjugates in All species samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen Mono- and polyubiquitinylated conjugates
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone UBCJ2
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias HEL-S-50, polyubiquitin B, polyubiquitin-B, UBB, UBC, ubiquitin B, ubiquitin C
Host Species Mouse
Immunogen Poly-ubiquitinylated lysozyme.
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7316
Target Species All species
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.