Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monocyte/Granulocyte Antibody (74-22-15), Biotin, Novus Biologicals™
SDP

Catalog No. NBP128243 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128243 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128243 Supplier Novus Biologicals Supplier No. NBP128243
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Monocyte/Granulocyte Monoclonal specifically detects Monocyte/Granulocyte in Mouse, Porcine samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunocytochemistry.

Specifications

Antigen Monocyte/Granulocyte
Applications Flow Cytometry, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone 74-22-15
Concentration 0.5mg/mL
Conjugate Biotin
Dilution Flow Cytometry <= 1 ug/10^6 cells, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry
Gene Alias 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor
Gene Symbols IL8
Host Species Mouse
Immunogen Fresh dd miniature swine thymocytes
Purification Method Protein A or G purified
Quantity 0.25 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3576
Test Specificity Porcine SWC3a
Target Species Mouse, Pig
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.