Learn More
Monocyte/Granulocyte Antibody (74-22-15), DyLight 680, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Monocyte/Granulocyte |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | 74-22-15 |
| Conjugate | DyLight 680 |
| Dilution | Flow Cytometry, ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Gene Alias | 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor |
| Gene Symbols | IL8 |
| Host Species | Mouse |
| Immunogen | Fresh dd miniature swine thymocytes |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.