Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monocyte/Macrophage Antibody (ER-HR3), Novus Biologicals™
SDP

Catalog No. NB119872 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119872 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119872 Supplier Novus Biologicals Supplier No. NBP311247
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Monocyte/Macrophage Monoclonal specifically detects Monocyte/Macrophage in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Monocyte/Macrophage
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone ER-HR3
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias macrophage, macrophages, monocyte, monocytes
Host Species Rat
Immunogen The monoclonal antibody ER-HR3 is reactive to the majority of mouse monocytes and a subset of mature resident macrophages, especially those located in hemopoietic organs.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2c
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.