Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monocyte/Macrophage Antibody (KUL01) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP128359 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128359 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128359 Supplier Novus Biologicals Supplier No. NBP128359
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Monocyte/Macrophage Monoclonal specifically detects Monocyte/Macrophage in Chicken samples. It is validated for Flow Cytometry.

Specifications

Antigen Monocyte/Macrophage
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone KUL01
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry <= 1 ug/10^6 cells
Gene Alias macrophage, macrophages, monocyte, monocytes
Host Species Mouse
Immunogen Chicken peripheral blood mononuclear cell leukocytes
Purification Method Protein A or G purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Chicken monocytes and macrophages
Target Species Chicken
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.