Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monomethyl Arginine Antibody (5D1), Novus Biologicals™
SDP

Catalog No. NB50012201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB50012201 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB50012201 Supplier Novus Biologicals Supplier No. NB5001220.1MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Monomethyl Arginine Monoclonal specifically detects Monomethyl Arginine in All Species samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Monomethyl Arginine
Applications ChIP Assay, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5D1
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, Chromatin Immunoprecipitation 1:10-1:500, Immunohistochemistry 1:100 - 1:10 000, Immunohistochemistry-Paraffin 1:100-1:10000
Gene Alias Arginine (mono methyl), Di methyl Arginine, Dimethyl Arginine, Methyl Arginine, Mono methyl Arginine, Monomethyl Arginine
Host Species Mouse
Immunogen Modified monomethyl arginine
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research
Primary or Secondary Primary
Test Specificity This reacts with free and bound NG-monomethyl arginine. There is no cross reaction with either NG-NG-dimethyl arginine or arginine.
Reconstitution Reconstitute with 0.1 ml distilled water.
Target Species All species
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.