Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monopolin Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB362184 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362184 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362184 Supplier Novus Biologicals Supplier No. NB500131MFV500

Rabbit Polyclonal Antibody

Monopolin Polyclonal antibody specifically detects Monopolin in Yeast samples. It is validated for Western Blot

Specifications

Antigen Monopolin
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a portion of human Monopolin.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Yeast
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.