Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOPS, 0.2M buffer soln., pH 6.0, low endotoxin, Thermo Scientific™

Catalog No. p-7057804
Click to view available options
Quantity:
100 mL

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 4-(3-sulfonatopropyl)morpholin-4-ium
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Color Colorless
Concentration 0.2 M
pH 6.0
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material MOPS, low endotoxin
Hazard Category H335
TSCA Yes
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.