Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOR-1C Antibody
SDP

Catalog No. NB11059977 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11059977 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11059977 Supplier Novus Biologicals Supplier No. NB11059977

Rabbit Polyclonal Antibody

MOR-1C Polyclonal specifically detects MOR-1C in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MOR-1C
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation Whole serum with 0.05% Sodium Azide
Gene Alias mu 1, mu opioid receptor mor1o; mor1; mu opiate receptor; kiaa0403; opioid receptor mu 1; mu opioid receptor; oprm; mutype opioid receptor; opioid receptor
Host Species Rabbit
Immunogen KSCMDRGMRNLLPDDGPRQE
Purification Method Protein A purified
Quantity 0.05 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity MOR-1C
Target Species Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.