Learn More
Description
Specifications
Specifications
| Antigen | MOR23-1 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:300 - 1:2000, Immunohistochemistry 1:300 - 1:2000, Immunohistochemistry-Paraffin 1:300 - 1:2000 |
| Formulation | Lyophilized from whole serum |
| Gene Alias | MOR23-1 olfactory receptor, Olfactory receptor OR11-33, Olfr599, OR11-33 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide from c-terminal region of mouse MOR23-1 conjugated to an immunogenic carrier protein was used as the antigen. |
| Purification Method | Unpurified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
