Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOR42-3 Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018027 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018027 0.1 mg
NB11075065 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018027 Supplier Novus Biologicals Supplier No. NB110750650.1MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MOR42-3 Polyclonal specifically detects MOR42-3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen MOR42-3
Applications Immunohistochemistry (Frozen), Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS with No Preservative
Gene Accession No. Q9WU88
Gene Alias Odorant receptor S6, Olfactory receptor 544, Olfactory receptor Olfr544, Olfr544., ORL463antibody, ORL471, S79
Host Species Rabbit
Immunogen A synthetic peptide as a part of mouse olfactory receptor family 42, subfamily 3 (MOR42-3) conjugated to immunogenic carrier protein was used as the immunogen.
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity MOR42-3.
Reconstitution Reconstitute with 0.5 ml sterilized water. Add glycerol (1:1) for additional stability.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.