Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MORC3 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB352826 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB352826 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB352826 Supplier Novus Biologicals Supplier No. NBP306255APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MORC3 Polyclonal antibody specifically detects MORC3 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen MORC3
Applications Immunocytochemistry
Classification Polyclonal
Conjugate APC
Dilution Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias KIAA0136, MORC family CW-type zinc finger 3, MORC family CW-type zinc finger protein 3, nuclear matrix protein NXP2, NXP2, ZCW5, ZCWCC3, Zinc finger CW-type coiled-coil domain protein 3, zinc finger, CW type with coiled-coil domain 3, zinc finger, CW-type with coiled-coil domain 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MORC3 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 23515
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.