Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MORC3 Mouse anti-Human,Mouse,Rat, Unconjugated, Clone: 17A9, MilliporeSigma™
Mouse Monoclonal Antibody
$189.29 - $481.52
Specifications
| Antigen | MORC3 |
|---|---|
| Clone | 17A9 |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MABC11052MI
|
MilliporeSigma
MABC110525UG |
25 μg |
Each for $189.29
|
|
|||||
|
MABC11051MI
|
MilliporeSigma
MABC1105100UG |
100 μg |
Each for $481.52
|
|
|||||
Description
MORC3 Monoclonal antibody specifically detects MORC3 in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Western Blotting.Specifications
| MORC3 | |
| Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| MORC family CW-type zinc finger protein 3;Nuclear matrix protein 2;Zinc finger CW-type coiled-coil domain protein 3 | |
| His-tagged full-length recombinant human MORC family CW-type zinc finger protein 3 (MORC3). | |
| Primary | |
| Stable for 1 year at 2 to 8°C from date of receipt. | |
| ∼160 and 230 kDa observed; 107.11 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
| 17A9 | |
| Monoclonal | |
| Purified | |
| Apoptosis and Cancer | |
| NP_056173 | |
| MORC3, KIAA0136, NXP2, ZCWCC3 | |
| IgG1 κ | |
| Protein G Purified | |
| Clone 17A9 is a mouse monoclonal antibody that specifically detects MORC family CW-type zinc finger protein 3 (MORC3). It targets an epitope with the C-terminal region. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title