Learn More
Description
Specifications
Specifications
| Antigen | MORN2 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 532 |
| Dilution | Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | BLOCK27, MOPT, MORN Motif Protein In Testis, MORN Repeat Containing 2, MORN Repeat-Containing Protein 2, Protein Containing Single MORN Motif In Testis |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human MORN2 fragment. |
| Purification Method | Antigen Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
