Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Morphine Antibody, Novus Biologicals™
SDP

Catalog No. NB10062764 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062764 0.25 mL
NB017508 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10062764 Supplier Novus Biologicals Supplier No. NB10062764
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Morphine Polyclonal specifically detects Morphine in All species samples. It is validated for ELISA.

Specifications

Antigen Morphine
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Formulation PBS with 0.09% Sodium Azide
Host Species Sheep
Immunogen Morphine conjugated to Keyhole Limpet Haemocyanin.
Purification Method Affinity Purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This product reacts with morphine and the related opiates: codeine, dihydrocodeine and morphine-3-glucuronide. Minimal reactivity has been observed with pethidine.
Target Species All species
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.