Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOS Antibody (R38.1), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB1002739AF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002739AF 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002739AF Supplier Novus Biologicals Supplier No. NB1002739AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MOS Monoclonal specifically detects MOS in Xenopus species samples. It is validated for ELISA, Western Blot.

Specifications

Antigen MOS
Applications ELISA, Western Blot
Classification Monoclonal
Clone R38.1
Conjugate Alexa Fluor 405
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias c-mos, EC 2.7.11, EC 2.7.11.1, MGC119962, MGC119963, MSV, oncogene MOS, Moloney murine sarcoma virus, Oocyte maturation factor mos, Proto-oncogene c-Mos, proto-oncogene serine/threonine-protein kinase mos, v-mos Moloney murine sarcoma viral oncogene homolog
Gene Symbols MOS
Host Species Mouse
Immunogen MBP-mos fusion protein.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 4342
Target Species Xenopus species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.