Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOS Antibody (S3.1) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB1002784 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002784 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002784 Supplier Novus Biologicals Supplier No. NB1002784
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MOS Monoclonal specifically detects MOS in Xenopus species samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen MOS
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone S3.1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500
Gene Alias c-mos, EC 2.7.11, EC 2.7.11.1, MGC119962, MGC119963, MSV, oncogene MOS, Moloney murine sarcoma virus, Oocyte maturation factor mos, Proto-oncogene c-Mos, proto-oncogene serine/threonine-protein kinase mos, v-mos Moloney murine sarcoma viral oncogene homolog
Gene Symbols MOS
Host Species Mouse
Immunogen Recombinant (Xenopus laevis) Mos fusion protein
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4342
Test Specificity Able to immunoprecipitate MOS from cell/egg extracts. Cross-reacts with Xenopus laevis.
Target Species Xenopus species
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.