Learn More
Description
Specifications
Specifications
| Antigen | MOT13 |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/Immunofluorescence 1-4 ug/ml |
| Formulation | PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide |
| Gene Alias | MCT 13, MCT13solute carrier family 16 (monocarboxylic acid transporters), member 13, monocarboxylate transporter 13, Solute carrier family 16 member 13, solute carrier family 16, member 13 (monocarboxylic acid transporter 13) |
| Gene Symbols | SLC16A13 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PPSLAEDPAVGGPRAQLTSLLHHGPF |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
