Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP11, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106488 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-488 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-488 Supplier Abnova Corporation Supplier No. H00011215A01
Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant AKAP11.

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein is expressed at high levels throughout spermatogenesis and in mature sperm. It binds the RI and RII subunits of PKA in testis. It may serve a function in cell cycle control of both somatic cells and germ cells in addition to its putative role in spermatogenesis and sperm function. [provided by RefSeq

Sequence: EGLGQDGKTLLITNIDMEPCTVDPQLRIILQWLIASEAEVAELYFHDSANKEFMLLSKQLQEKGWKVGDLLQAVLQYYEVMEKASSEERCKSLFDWLLENA

Specifications

Antigen AKAP11
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant AKAP11.
Formulation 50% glycerol
Gene AKAP11
Gene Accession No. NM_016248
Gene Alias AKAP220/DKFZp781I12161/FLJ11304/KIAA0629/PRKA11
Gene Symbols AKAP11
Host Species Mouse
Immunogen AKAP11 (NP_057332, 1801 a.a. to 1901 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11215
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.