Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-Fetoprotein/AFP Antibody (MBS-12), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
| Antigen | alpha-Fetoprotein/AFP |
|---|---|
| Clone | MBS-12 |
| Concentration | 0.2mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24501300
![]() |
Novus Biologicals
NBP2450130.02MG |
0.02 mg |
Each for $268.00
|
|
|||||
NBP24501301
![]() |
Novus Biologicals
NBP2450130.1MG |
0.1 mg |
Each for $524.00
|
|
|||||
NBP24501302
![]() |
Novus Biologicals
NBP2450130.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
alpha-Fetoprotein/AFP Monoclonal specifically detects alpha-Fetoprotein/AFP in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| alpha-Fetoprotein/AFP | |
| 0.2mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Cancer Stem Cells, GPCR, Stem Cell Markers, Tumor Biomarkers | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP | |
| AFP | |
| IgG1 κ | |
| Protein A or G purified | |
| It recognizes an oncofetal glycoprotein with a single chain of 70kDa, which is identified as alpha fetoprotein (AFP). This MAb is highly specific to AFP and shows no cross-reaction with other oncofetal antigens or serum albumin. The yolk sac and the liver produce AFP during fetal life. AFP expression in adults is often associated with hepatoma or teratoma. However, hereditary persistence of alpha-fetoprotein may also be found in individuals with no obvious pathology. The protein is thought to be the fetal counterpart of serum albumin, and the AFP and albumin genes are present in tandem in the same transcriptional orientation on chromosome 4. AFP is found in monomeric as well as dimeric and trimeric forms, and binds copper, nickel, fatty acids and bilirubin. The level of AFP in amniotic fluid is used to measure renal loss of protein to screen for spinal bifida and anencephaly. |
| MBS-12 | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P02771 | |
| 174 | |
| Recombinant full-length human Alpha fetoprotein | |
| Primary | |
| Store at 4C. | |
| 70 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title