Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP1S2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105976 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-976 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-976 Supplier Abnova Corporation Supplier No. H00008905A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant AP1S2.

Adaptor protein complex 1 is found at the cytoplasmic face of coated vesicles located at the Golgi complex, where it mediates both the recruitment of clathrin to the membrane and the recognition of sorting signals within the cytosolic tails of transmembrane receptors. This complex is a heterotetramer composed of two large, one medium, and one small adaptin subunit. The protein encoded by this gene serves as the small subunit of this complex and is a member of the adaptin protein family. Transcript variants utilizing alternative polyadenylation signals exist for this gene. [provided by RefSeq

Sequence: MQFMLLFSRQGKLRLQKWYVPLSDKEKKKITRELVQTVLARKPKMCSFLEWRDLKIVYKRYASLYFCCAIEDQDNELITLEIIHRYVELLDKYFGSVCELDIIFNFEKAYFILDEFLLGGEVQETSKKNVLKAIEQADLLQEEAETPRSVLEEIGLT

Specifications

Antigen AP1S2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant AP1S2.
Formulation 50% glycerol
Gene AP1S2
Gene Accession No. BC001117
Gene Alias DC22/MGC:1902/MRX59/SIGMA1B
Gene Symbols AP1S2
Host Species Mouse
Immunogen AP1S2 (AAH01117, 1 a.a. to 157 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8905
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.