Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-BMP-4, Clone: 26H16, Novus Biologicals™
SDP

Catalog No. NBP11920705 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP11920705 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP11920705 Supplier Novus Biologicals Supplier No. NBP1192070.05MG

Mouse Monoclonal Antibody

BMP-4 Monoclonal specifically detects BMP-4 in Human samples. It is validated for Western Blot.

Specifications

Antigen BMP-4
Applications Western Blot
Classification Monoclonal
Clone 26H16
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml
Formulation PBS (pH 7.2), BSA and 50% Glycerol with 0.09% Sodium Azide
Gene Accession No. P12644
Gene Alias BMP-2B, BMP2B1, BMP2BMCOPS6, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, OFC11, ZYME
Gene Symbols BMP4
Host Species Mouse
Immunogen Recombinant full-length human BMP4.
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Embryonic Stem Cell Markers, Stem Cell Markers, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 652
Reconstitution Protein A purified
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.