Learn More
Mouse anti-CCL16/HCC-4/LEC, Clone: LEC66, Azide Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | CCL16/HCC-4/LEC |
| Applications | ELISA, Functional Assay |
| Classification | Monoclonal |
| Clone | LEC66 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | ELISA 1-5ug/ml for coating, Functional |
| Formulation | 10mM PBS - BSA free with No Preservative |
| Gene Accession No. | O15467 |
| Gene Alias | C-C motif chemokine 16, chemokine (C-C motif) ligand 16, Chemokine CC-4, Chemokine LEC, CKb12, HCC-4Liver-expressed chemokine, LCC-1Lymphocyte and monocyte chemoattractant, LEC, liver CC chemokine-1, LMCSmall-inducible cytokine A16, monotactin-1, MTN-1, NCC4IL-10-inducible chemokine, NCC-4ILINCK, new CC chemokine 4, SCYA16MGC117051, SCYL4, small inducible cytokine subfamily A (Cys-Cys), member 16 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.