Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-CD163, Clone: RM3/1, Novus Biologicals™
SDP

Catalog No. NBP16015805 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP16015805 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP16015805 Supplier Novus Biologicals Supplier No. NBP1601580.05MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 5 publications

CD163 Monoclonal specifically detects CD163 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.

Specifications

Antigen CD163
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone RM3/1
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:10, Flow Cytometry 1:10, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10
Formulation 0.2 um filtered solution in PBS and 0.1% BSA with 0.02% Sodium Azide
Gene Alias CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130
Gene Symbols CD163
Host Species Mouse
Immunogen Human monocytes (epitope recognized SRCR domain 9)
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9332
Test Specificity Guinea Pig (Negative), Porcine (Negative), Rat (Negative)
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.