Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-CD2, Clone: BH1, Azide Free, Novus Biologicals™
SDP

Catalog No. NBP23456102 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP23456102 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP23456102 Supplier Novus Biologicals Supplier No. NBP2345610.2 MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

CD2 Monoclonal antibody specifically detects CD2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Functional, CyTOF-ready.

Specifications

Antigen CD2
Applications Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone BH1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5-1ug/million cells, ELISA 1-5ug/ml for coating, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Frozen 0.5-1ug/ml, Functional, CyTOF-ready
Formulation 10mM PBS - BSA free with No Preservative
Gene Accession No. P06729
Gene Alias CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5
Gene Symbols CD2
Host Species Mouse
Immunogen Human CD2 protein
Molecular Weight of Antigen 50 kDa
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 914
Test Specificity CD2 interacts through its amino-terminal domain with the extracellular domain of CD58 (also designated CD2 ligand) to mediate cell adhesion. CD2/CD58 binding can enhance antigen-specific T cell activation. CD2 is a transmembrane glycoprotein that is expressed on peripheral blood T lymphocytes, NK cells and thymocytes. CD58 is a heavily glycosylated protein with a broad tissue distribution in hematopoietic and other cells, including endothelium. Interaction between CD2 and its counter receptor LFA3 (CD58) on opposing cells optimizes immune system recognition, thereby facilitating communication between helper T lymphocytes and antigen-presenting cells, as well as between cytolytic effectors and target cells.
Reconstitution Protein A purified
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.