Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-CD20, Clone: 2H7, PE,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABF1636
Description
Specifically detects CD20 clone: 2H7 in Human samples, and it is validated for Flow Cytometry
The 2H7 antibody reacts with human CD20, expressed by pre-B cells, resting and activated B cells, follicular dendritic cells and a small subset of T cells, but not on plasma cells. CD20 is a 33-37kDa transmembrane protein which is differentially phosphorylated in resting and active B cells. The CD20 molecule plays a role in regulating B-cell activation.The 2H7 antibody is reported to be cross-reactive with several non-human species including Baboon, Chimpanzee, Cynomolgus and Rhesus.
Specifications
| CD20 | |
| Monoclonal | |
| PE | |
| P11836 | |
| Mouse | |
| Purified by size-exclusion chromatography | |
| RUO | |
| Primary | |
| Stable for one year at 2°C to 8°C from date of receipt. Note: It is recommended to store the product undiluted at 2°C to 8°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 2H7 | |
| Purified Mouse monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
| MS4A1; CD20 | |
| Immunogen information for this antibody is proprietary information. | |
| 100 Tests | |
| Inflammation and Immunology | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction