Learn More
CD25 Mouse anti-Human, APC-R700, Clone: 2A3, BD
Mouse Monoclonal Antibody
$178.80 - $378.00
Specifications
Antigen | CD25 |
---|---|
Clone | 2A3 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC-R700 |
Description
The 2A3 monoclonal antibody specifically binds to human CD25, the low-affinity alpha subunit of the Interleukin-2 Receptor (IL- 2Rα). CD25 associates with CD122 (IL-2Rβ chain) and CD132 (common γchain or γc) to form the high-affinity signal-transducing IL -2R complex. CD25 is expressed by subsets of thymocytes and peripheral blood lymphocytes including CD4+CD25+ regulatory T cells and memory T cells. CD25 antigen density increases on activated T cells including phytohemagglutinin (PHA)-, concanavalin A (Con A)-, and CD3-activated T lymphocytes. High levels of CD25 can be expressed by T lymphocytes from mixed lymphocyte cultures and by human T-lymphocyte leukemia virus (HTLV)-infected T-lymphocyte leukemia lines, for example, HUT-102. CD25 can also be expressed by activated B cells and macrophages. Recombinant IL-2 blocks the binding of the 2A3 antibody to PHA-activated T lymphocytes.
This antibody was conjugated to BD Horizon APC-R700, which has been developed exclusively by BD Biosciences as a better alternative to Alexa Fluor™ 700. APC-R700 excites and emits at similar wavelengths to Alexa Fluor™ 700 yet exhibits significantly improved brightness. This dye can be excited by the red laser and detected with the same filter set as Alexa Fluor™ (eg, 730/45-nm filter).
Specifications
CD25 | |
Flow Cytometry | |
APC-R700 | |
RUO | |
100 | |
IgG1 κ | |
Affinity Purified | |
IL-2R; IL2RA; IL-2Rα; TCGFR; TAC antigen; p55 |
2A3 | |
Monoclonal | |
Mouse | |
Human | |
Human Phytohemagglutinin-activated T Cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.