Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-CD38, Clone: HIT2, PE,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABF1626
Description
Specifically detects CD38 clone: HIT2 in Human samples, and it is validated for Flow Cytometry
The HIT2 antibody is specific for human CD38, a 45 kD type II transmembrane glycoprotein expressed on thymocytes, activated T and B cells, and monocytes as well as other non-hematopoietic cells. CD38 is an ectoenzyme that functions to catalyze the synthesis and hydrolysis of cyclic ADP-ribose and is involved in cell signaling, regulating cell adhesion, and activation. Additionally, CD38 has been shown to be a prognostic marker for some leukemias and other diseases.
Specifications
CD38 | |
Monoclonal | |
PE | |
P28907 | |
Mouse | |
Purified by size-exclusion chromatography | |
RUO | |
Primary | |
Stable for one year at 2°C to 8°C from date of receipt. Note: It is recommended to store the product undiluted at 2°C to 8°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
HIT2 | |
Purified Mouse monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
CD38 | |
Immunogen information for this antibody is proprietary information. | |
100 Tests | |
Inflammation and Immunology | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction