Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-CD38, Clone: HIT2, PE,
Mouse Monoclonal Antibody
$144.24 - $321.52
Specifications
Antigen | CD38 |
---|---|
Clone | HIT2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Description
Specifically detects CD38 clone: HIT2 in Human samples, and it is validated for Flow Cytometry
The HIT2 antibody is specific for human CD38, a 45 kD type II transmembrane glycoprotein expressed on thymocytes, activated T and B cells, and monocytes as well as other non-hematopoietic cells. CD38 is an ectoenzyme that functions to catalyze the synthesis and hydrolysis of cyclic ADP-ribose and is involved in cell signaling, regulating cell adhesion, and activation. Additionally, CD38 has been shown to be a prognostic marker for some leukemias and other diseases.Specifications
CD38 | |
Flow Cytometry | |
PE | |
Mouse | |
Inflammation and Immunology | |
Purified Mouse monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
CD38 | |
Primary | |
Stable for one year at 2°C to 8°C from date of receipt. Note: It is recommended to store the product undiluted at 2°C to 8°C and protected from prolonged exposure to light. Do not freeze. |
HIT2 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P28907 | |
Immunogen information for this antibody is proprietary information. | |
Purified by size-exclusion chromatography |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title